siRNA Details
| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081428.1 | NP_001074897.1 | 2 | 484 | ||
| GenBank mRNA | AK015334.1 | - | 2 | 485 | ||
| EF410115.1 | - | 541 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam184a| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081428.1 | NP_001074897.1 | 2 | 484 | ||
| GenBank mRNA | AK015334.1 | - | 2 | 485 | ||
| EF410115.1 | - | 541 | ||||