siRNA Details
| Gene Symbol: 4930579G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029482.1 | NP_083758.1 | 2 | 1681 | ||
| GenBank mRNA | AK137776.1 | - | 1670 | |||
| BC052483.1 | - | 2 | 1662 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930579G24Rik| Gene Symbol: 4930579G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029482.1 | NP_083758.1 | 2 | 1681 | ||
| GenBank mRNA | AK137776.1 | - | 1670 | |||
| BC052483.1 | - | 2 | 1662 | |||