siRNA Details
| Gene Symbol: Usp31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033173.1 | NP_001028345.1 | 16 | 5338 | ||
| GenBank mRNA | AK018244.1 | - | 1 | 24 | ||
| AK122466.1 | - | 1 | 2158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp31| Gene Symbol: Usp31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033173.1 | NP_001028345.1 | 16 | 5338 | ||
| GenBank mRNA | AK018244.1 | - | 1 | 24 | ||
| AK122466.1 | - | 1 | 2158 | |||