siRNA Details
| Gene Symbol: Gpr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029536.3 | NP_083812.1 | 3 | 1230 | ||
| GenBank mRNA | AK018327.1 | - | 3 | 1258 | ||
| AK034479.1 | - | 3 | 1174 | |||
| AK038566.1 | - | 3 | 502 | |||
| AK042723.1 | - | 3 | 1207 | |||
| BC113184.1 | - | 3 | 1081 | 40061761 | ||
| BC113791.1 | - | 3 | 1081 | 40061764 | ||
More Information
Set Membership: |
|

Gpr165
Genomic Map