siRNA Details
| Gene Symbol: Gpr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029536.3 | NP_083812.1 | 5 | 2716 | ||
| GenBank mRNA | AK034479.1 | - | 5 | 2660 | ||
| AK038566.1 | - | 4 | 2593 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr165| Gene Symbol: Gpr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029536.3 | NP_083812.1 | 5 | 2716 | ||
| GenBank mRNA | AK034479.1 | - | 5 | 2660 | ||
| AK038566.1 | - | 4 | 2593 | |||
Set Membership: |
|