siRNA Details
| Gene Symbol: Gpr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029536.3 | NP_083812.1 | 5 | 3166 | ||
| GenBank mRNA | AK034479.1 | - | 5 | 3110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr165| Gene Symbol: Gpr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029536.3 | NP_083812.1 | 5 | 3166 | ||
| GenBank mRNA | AK034479.1 | - | 5 | 3110 | ||
Set Membership: |
|