siRNA Details
| Gene Symbol: Jakmip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163637.1 | NP_001157109.1 | 6, 7 | 1547 | ||
| GenBank mRNA | AK133750.1 | - | 6, 7 | 1512 | ||
| AK139194.1 | - | 1, 2 | 2390 | |||
| BC094439.1 | - | 5, 6 | 1108 | |||

Jakmip2
Genomic Map