siRNA Details
Gene Symbol: Btg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019290.1 | NP_062163.1 | 4 | 424 | ||
GenBank mRNA | AF087037.1 | - | 4 | 424 | ||
BC127498.1 | - | 4 | 517 | 8371953 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019290.1 | NP_062163.1 | 4 | 424 | ||
GenBank mRNA | AF087037.1 | - | 4 | 424 | ||
BC127498.1 | - | 4 | 517 | 8371953 |