siRNA Details
Gene Symbol: Zfp773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029584.1 | NP_083860.1 | 4 | 1999 | ||
GenBank mRNA | AK013056.1 | - | 4 | 1967 | ||
AK031040.1 | - | 4 | 1999 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029584.1 | NP_083860.1 | 4 | 1999 | ||
GenBank mRNA | AK013056.1 | - | 4 | 1967 | ||
AK031040.1 | - | 4 | 1999 |