siRNA Details
| Gene Symbol: 1700019B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029598.1 | NP_083874.1 | 9 | 1594 | ||
| GenBank mRNA | AK006106.1 | - | 6 | 839 | ||
| BC050809.1 | - | 9 | 1589 | 6771839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700019B03Rik| Gene Symbol: 1700019B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029598.1 | NP_083874.1 | 9 | 1594 | ||
| GenBank mRNA | AK006106.1 | - | 6 | 839 | ||
| BC050809.1 | - | 9 | 1589 | 6771839 | ||