siRNA Details
| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146710.1 | NP_001140182.1 | 4 | 2621 | ||
| NM_001146711.1 | NP_001140183.1 | 4 | 2620 | |||
| NM_175242.1 | NP_780451.1 | 3 | 3135 | |||
| GenBank mRNA | AK009340.1 | - | 3 | 1109 | ||
| AK051656.1 | - | 3 | 3090 | |||
| AK154854.1 | - | 3 | 1348 | |||
| BC025573.1 | - | 3 | 3492 | |||

Ppp1r18
Genomic Map