siRNA Details
| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146710.1 | NP_001140182.1 | 2 | 837 | ||
| NM_001146711.1 | NP_001140183.1 | 2 | 836 | |||
| NM_175242.1 | NP_780451.1 | 1 | 1351 | |||
| GenBank mRNA | AK051656.1 | - | 1 | 1307 | ||
| AK149943.1 | - | 2 | 838 | |||
| BC020483.1 | - | 2 | 909 | |||
| BC025573.1 | - | 2 | 849 | |||
| BC145183.1 | - | 2 | 806 | 9053734 | ||
| BC150725.1 | - | 2 | 806 | 9087636 | ||

Ppp1r18
Genomic Map