siRNA Details
Gene Symbol: Ccdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111121.1 | NP_001104591.1 | 9 | 5149 | ||
GenBank mRNA | AK012728.1 | - | 1 | 814 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111121.1 | NP_001104591.1 | 9 | 5149 | ||
GenBank mRNA | AK012728.1 | - | 1 | 814 |