siRNA Details
| Gene Symbol: Snx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190156.1 | NP_001177085.1 | 8 | 1299 | ||
| NM_029655.3 | NP_083931.2 | 9 | 1452 | |||
| GenBank mRNA | AK011015.1 | - | 9 | 1427 | ||
| AK038971.1 | - | 9 | 1453 | |||
| AK170966.1 | - | 8 | 1246 | |||
| BC087942.1 | - | 9 | 1310 | 30303462 | ||

Snx7
Genomic Map