siRNA Details
| Gene Symbol: Prr30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029680.1 | NP_083956.1 | 2 | 1837 | ||
| GenBank mRNA | AK007164.1 | - | 1 | 1688 | ||
| BX521393.1 | - | 1 | 329 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr30| Gene Symbol: Prr30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029680.1 | NP_083956.1 | 2 | 1837 | ||
| GenBank mRNA | AK007164.1 | - | 1 | 1688 | ||
| BX521393.1 | - | 1 | 329 | |||