siRNA Details
| Gene Symbol: 1700112E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028275.1 | NP_082551.1 | 7 | 840 | ||
| GenBank mRNA | AK007175.1 | - | 7 | 844 | ||
| AK153491.1 | - | 6 | 675 | |||
| BY516672.1 | - | 355 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700112E06Rik| Gene Symbol: 1700112E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028275.1 | NP_082551.1 | 7 | 840 | ||
| GenBank mRNA | AK007175.1 | - | 7 | 844 | ||
| AK153491.1 | - | 6 | 675 | |||
| BY516672.1 | - | 355 | ||||