siRNA Details
| Gene Symbol: 1700122O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029689.1 | NP_083965.1 | 3 | 738 | ||
| GenBank mRNA | AK007241.1 | - | 3 | 742 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700122O11Rik| Gene Symbol: 1700122O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029689.1 | NP_083965.1 | 3 | 738 | ||
| GenBank mRNA | AK007241.1 | - | 3 | 742 | ||