siRNA Details
| Gene Symbol: Arpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029711.1 | NP_083987.1 | 3 | 432 | ||
| GenBank mRNA | AK083263.1 | - | 3 | 242 | ||
| AK151513.1 | - | 3 | 406 | |||
| AK152356.1 | - | 4 | 257 | |||
| AK170061.1 | - | 4 | 231 | |||
| BC099504.1 | - | 2 | 84 | |||
| BC115750.1 | - | 3 | 230 | 40051600 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106919, XM_006245166, XM_006245165, FQ231947, FQ214133.

Arpc2
Genomic Map