siRNA Details
| Gene Symbol: 1700039E15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033176.1 | NP_001028348.1 | 6 | 833 | ||
| GenBank mRNA | AK133083.1 | - | 6 | 833 | ||
| BC100502.1 | - | 2 | 635 | |||
| BC147591.1 | - | 6 | 930 | 9056581 | ||
| BC147593.1 | - | 6 | 930 | 9056583 | ||
 Back To Top
 Back To Top
 1700039E15Rik
1700039E15Rik Genomic Map
Genomic Map Back To Top
 Back To Top