siRNA Details
| Gene Symbol: 1700039E15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033176.1 | NP_001028348.1 | 6 | 1705 | ||
| GenBank mRNA | AK133083.1 | - | 6 | 1705 | ||
| BC100502.1 | - | 2 | 1507 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700039E15Rik| Gene Symbol: 1700039E15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033176.1 | NP_001028348.1 | 6 | 1705 | ||
| GenBank mRNA | AK133083.1 | - | 6 | 1705 | ||
| BC100502.1 | - | 2 | 1507 | |||