siRNA Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081045.1 | NP_001074514.1 | 14 | 4784 | ||
| GenBank mRNA | AK122484.1 | - | 14 | 4790 | ||
| AK161032.1 | - | 3 | 2556 | |||
| BC025052.1 | - | 4 | 1665 | |||
| BC039751.1 | - | 1 | 273 | |||
| BC043121.1 | - | 14 | 4333 | 6409084 | ||
| BC054752.1 | - | 15 | 4607 | 6834541 | ||
| BC079594.1 | - | 14 | 4332 | 6827379 | ||

Kansl1
Genomic Map