siRNA Details
| Gene Symbol: Snx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082484.2 | NP_001075953.1 | 12 | 3015 | ||
| NM_029721.2 | NP_083997.1 | 12 | 3809 | |||
| GenBank mRNA | AK043074.1 | - | 3 | 1399 | ||
| AK122296.1 | - | 12 | 3621 | |||
| AK147452.1 | - | 12 | 3795 | |||
| AK164500.1 | - | 1 | 1244 | |||
| AK167610.1 | - | 1 | 953 | |||

Snx27
Genomic Map