siRNA Details
| Gene Symbol: Gje1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029722.1 | NP_083998.1 | 3 | 607 | ||
| GenBank mRNA | AK018698.1 | - | 3 | 608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gje1| Gene Symbol: Gje1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029722.1 | NP_083998.1 | 3 | 607 | ||
| GenBank mRNA | AK018698.1 | - | 3 | 608 | ||