siRNA Details
Gene Symbol: 2410124H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029740.1 | NP_084016.1 | 3 | 533 | ||
GenBank mRNA | AK010774.1 | - | 3 | 534 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2410124H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029740.1 | NP_084016.1 | 3 | 533 | ||
GenBank mRNA | AK010774.1 | - | 3 | 534 |