siRNA Details
| Gene Symbol: Nubpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029760.2 | NP_084036.2 | 9 | 834 | ||
| GenBank mRNA | AK010827.1 | - | 9 | 816 | ||
| AK141257.1 | - | 9 | 798 | |||
| AK152377.1 | - | 9 | 835 | |||
| BC023294.1 | - | 428 | ||||
| BC062175.1 | - | 9 | 815 | 30291107 | ||

Nubpl
Genomic Map