siRNA Details
| Gene Symbol: Morn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081100.1 | NP_001074569.1 | 14 | 1631 | ||
| GenBank mRNA | AK045316.1 | - | 14 | 1606 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morn1| Gene Symbol: Morn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081100.1 | NP_001074569.1 | 14 | 1631 | ||
| GenBank mRNA | AK045316.1 | - | 14 | 1606 | ||