siRNA Details
| Gene Symbol: Tstd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029840.1 | NP_084116.1 | 4 | 1157 | ||
| GenBank mRNA | AK011611.1 | - | 4 | 1072 | ||
| AK154853.1 | - | 4 | 1144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tstd3| Gene Symbol: Tstd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029840.1 | NP_084116.1 | 4 | 1157 | ||
| GenBank mRNA | AK011611.1 | - | 4 | 1072 | ||
| AK154853.1 | - | 4 | 1144 | |||