siRNA Details
| Gene Symbol: Krt76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033177.2 | NP_001028349.1 | 9 | 2076 | ||
| GenBank mRNA | AK009075.1 | - | 9 | 2060 | ||
| AK036802.1 | - | 9 | 2077 | |||
| AK137676.1 | - | 9 | 2077 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt76| Gene Symbol: Krt76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033177.2 | NP_001028349.1 | 9 | 2076 | ||
| GenBank mRNA | AK009075.1 | - | 9 | 2060 | ||
| AK036802.1 | - | 9 | 2077 | |||
| AK137676.1 | - | 9 | 2077 | |||