siRNA Details
| Gene Symbol: Ston1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029858.2 | NP_084134.2 | 3 | 2340 | ||
| GenBank mRNA | AK014958.1 | - | 3 | 2275 | ||
| AK029878.1 | - | 2287 | ||||
| AK029959.1 | - | 2320 | ||||
| CF729405.1 | - | 2 | 544 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ston1| Gene Symbol: Ston1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029858.2 | NP_084134.2 | 3 | 2340 | ||
| GenBank mRNA | AK014958.1 | - | 3 | 2275 | ||
| AK029878.1 | - | 2287 | ||||
| AK029959.1 | - | 2320 | ||||
| CF729405.1 | - | 2 | 544 | |||