siRNA Details
| Gene Symbol: Hnrnpa0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029872.1 | NP_084148.1 | 1 | 1882 | ||
| GenBank mRNA | AK004260.1 | - | 1 | 189 | ||
| AK172180.1 | - | 1 | 395 | |||
| BC023150.1 | - | 1 | 713 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpa0| Gene Symbol: Hnrnpa0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029872.1 | NP_084148.1 | 1 | 1882 | ||
| GenBank mRNA | AK004260.1 | - | 1 | 189 | ||
| AK172180.1 | - | 1 | 395 | |||
| BC023150.1 | - | 1 | 713 | |||