siRNA Details
| Gene Symbol: Krtap5-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023860.1 | NP_076349.1 | 1 | 51 | ||
| GenBank mRNA | AK020699.1 | - | 1 | 52 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap5-3| Gene Symbol: Krtap5-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023860.1 | NP_076349.1 | 1 | 51 | ||
| GenBank mRNA | AK020699.1 | - | 1 | 52 | ||