siRNA Details
Gene Symbol: Fam162b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029894.1 | NP_084170.1 | 4 | 429 | ||
GenBank mRNA | AK020485.1 | - | 4 | 450 | ||
BC147085.1 | - | 4 | 429 | 9056123 | ||
BC147086.1 | - | 4 | 429 | 9056133 |
