siRNA Details
| Gene Symbol: Raph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045513.3 | NP_001038978.3 | 14 | 7697 | ||
| GenBank mRNA | AK034402.1 | - | 1 | 911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Raph1| Gene Symbol: Raph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045513.3 | NP_001038978.3 | 14 | 7697 | ||
| GenBank mRNA | AK034402.1 | - | 1 | 911 | ||