siRNA Details
| Gene Symbol: Madcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019317.1 | NP_062190.1 | 3, 4 | 659 | ||
| XM_008765179.1 | XP_008763401.1 | 3, 4 | 971 | |||
| GenBank mRNA | D87840.1 | - | 3, 4 | 659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Madcam1| Gene Symbol: Madcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019317.1 | NP_062190.1 | 3, 4 | 659 | ||
| XM_008765179.1 | XP_008763401.1 | 3, 4 | 971 | |||
| GenBank mRNA | D87840.1 | - | 3, 4 | 659 | ||