siRNA Details
| Gene Symbol: Dcst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030269.1 | - | 10 | 1406 | ||
| BC026583.1 | - | 2 | 182 | |||
| BC125478.1 | - | 10 | 1391 | 40129993 | ||
| BC125480.1 | - | 10 | 1391 | 40129995 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcst1| Gene Symbol: Dcst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030269.1 | - | 10 | 1406 | ||
| BC026583.1 | - | 2 | 182 | |||
| BC125478.1 | - | 10 | 1391 | 40129993 | ||
| BC125480.1 | - | 10 | 1391 | 40129995 | ||