siRNA Details
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100451.2 | NP_001093921.1 | 4829 | |||
GenBank mRNA | AK035934.1 | - | 1 | 3049 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100451.2 | NP_001093921.1 | 4829 | |||
GenBank mRNA | AK035934.1 | - | 1 | 3049 |