siRNA Details
| Gene Symbol: Krtap17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099774.2 | NP_001093244.1 | 1 | 382 | ||
| GenBank mRNA | AK020698.1 | - | 1 | 383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap17-1| Gene Symbol: Krtap17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099774.2 | NP_001093244.1 | 1 | 382 | ||
| GenBank mRNA | AK020698.1 | - | 1 | 383 | ||