siRNA Details
Gene Symbol: 9230112D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030062.1 | NP_084338.1 | 3 | 657 | ||
GenBank mRNA | AK020336.1 | - | 3 | 658 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9230112D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030062.1 | NP_084338.1 | 3 | 657 | ||
GenBank mRNA | AK020336.1 | - | 3 | 658 |