siRNA Details
| Gene Symbol: 9230112D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030062.1 | NP_084338.1 | 3 | 464 | ||
| GenBank mRNA | AK020336.1 | - | 3 | 465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230112D13Rik| Gene Symbol: 9230112D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030062.1 | NP_084338.1 | 3 | 464 | ||
| GenBank mRNA | AK020336.1 | - | 3 | 465 | ||