siRNA Details
| Gene Symbol: Adgrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289499.1 | NP_001276428.1 | 2 | 195 | ||
| NM_001289500.1 | NP_001276429.1 | 2 | 182 | |||
| NM_001289501.1 | NP_001276430.1 | 2 | 226 | |||
| NM_030067.2 | NP_084343.1 | 2 | 269 | |||
| GenBank mRNA | AK019508.1 | - | 2 | 272 | ||
| BC089564.1 | - | 2 | 195 | 30502915 | ||
| BY142177.1 | - | 2 | 217 | |||
| BY714064.1 | - | 2 | 270 | |||

Adgrf4
Genomic Map