siRNA Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 7 | 1816 | ||
| GenBank mRNA | AK122351.1 | - | 4 | 807 | ||
| AK147675.1 | - | 7 | 2264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul9| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 7 | 1816 | ||
| GenBank mRNA | AK122351.1 | - | 4 | 807 | ||
| AK147675.1 | - | 7 | 2264 | |||