siRNA Details
| Gene Symbol: Padi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019332.1 | NP_062205.1 | 2 | 279 | ||
| GenBank mRNA | AB010998.1 | - | 2 | 279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi1| Gene Symbol: Padi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019332.1 | NP_062205.1 | 2 | 279 | ||
| GenBank mRNA | AB010998.1 | - | 2 | 279 | ||