siRNA Details
| Gene Symbol: Lce6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166172.1 | NP_001159644.1 | 3 | 429 | ||
| GenBank mRNA | AK019072.1 | - | 2 | 125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lce6a| Gene Symbol: Lce6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166172.1 | NP_001159644.1 | 3 | 429 | ||
| GenBank mRNA | AK019072.1 | - | 2 | 125 | ||