siRNA Details
| Gene Symbol: 1700057G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033184.3 | NP_001028356.1 | 7 | 945 | ||
| GenBank mRNA | AK132905.1 | - | 7 | 946 | ||
| BC147543.1 | - | 6 | 835 | 9056534 | ||
| BC172079.1 | - | 5 | 792 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700057G04Rik| Gene Symbol: 1700057G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033184.3 | NP_001028356.1 | 7 | 945 | ||
| GenBank mRNA | AK132905.1 | - | 7 | 946 | ||
| BC147543.1 | - | 6 | 835 | 9056534 | ||
| BC172079.1 | - | 5 | 792 | |||