siRNA Details
| Gene Symbol: 1700084C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033185.2 | NP_001028357.1 | 4 | 915 | ||
| GenBank mRNA | AK141447.1 | - | 4 | 916 | ||
| BC172001.1 | - | 3 | 843 | 9054287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700084C01Rik| Gene Symbol: 1700084C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033185.2 | NP_001028357.1 | 4 | 915 | ||
| GenBank mRNA | AK141447.1 | - | 4 | 916 | ||
| BC172001.1 | - | 3 | 843 | 9054287 | ||