siRNA Details
| Gene Symbol: Pitx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042505.1 | NP_001035970.1 | 2 | 581 | ||
| NM_019334.2 | NP_062207.1 | 3 | 282 | |||
| GenBank mRNA | AF039832.1 | - | 3 | 282 | ||
| AJ222971.1 | - | 2 | 589 | |||
| AJ222972.1 | - | 3 | 335 | |||
| EF519321.1 | - | 2 | 71 | |||

Pitx2
Genomic Map