siRNA Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191004.1 | NP_001177933.1 | 5 | 916 | ||
| NM_030145.3 | NP_084421.1 | 4 | 817 | |||
| GenBank mRNA | AK078392.1 | - | 5 | 866 | ||
| AK141884.1 | - | 5 | 863 | |||
| AK158236.1 | - | 4 | 806 | |||
| AK161686.1 | - | 5 | 870 | |||
| BC030427.1 | - | 4 | 768 | 5370660 | ||
| BC098230.1 | - | 4 | 767 | 3372313 | ||

Lsm6
Genomic Map