siRNA Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191004.1 | NP_001177933.1 | 5 | 2772 | ||
| NM_030145.3 | NP_084421.1 | 4 | 2673 | |||
| GenBank mRNA | AK141884.1 | - | 5 | 2719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm6| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191004.1 | NP_001177933.1 | 5 | 2772 | ||
| NM_030145.3 | NP_084421.1 | 4 | 2673 | |||
| GenBank mRNA | AK141884.1 | - | 5 | 2719 | ||