siRNA Details
| Gene Symbol: Ltn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081068.1 | NP_001074537.1 | 29 | 5253 | ||
| GenBank mRNA | AK173009.1 | - | 29 | 5124 | ||
| BC027795.1 | - | 8 | 1223 | |||
| BC030355.1 | - | 4 | 594 | |||
| BC039795.1 | - | 2 | 574 | |||

Ltn1
Genomic Map