siRNA Details
| Gene Symbol: Saal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030233.1 | NP_084509.1 | 12 | 4535 | ||
| GenBank mRNA | AK132193.1 | - | 12 | 4536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saal1| Gene Symbol: Saal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030233.1 | NP_084509.1 | 12 | 4535 | ||
| GenBank mRNA | AK132193.1 | - | 12 | 4536 | ||